Bromobenzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
Phenylbromide, Phenyl bromide
221285-10KG
221285-2.5KG
221285-500G
BT221285-10KGEA
367
USD
BT221285-10KG
BT221285-2.5KG
BT221285-500G
Bromobenzene ≥99%
Bromobenzene
CAS: 108-86-1; EC No: 203-623-8; MDL No: MFCD00000055; RTECS: CY9000000
UN No: UN2514; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H5Br; MW: 157.01
Melting Point: -31°; Boiling Point: 156°; Flash point: 51°C (124°F)
Density (g/mL): 1.491; Refractive Index: 1.559
- RTECS: CY9000000
- Refractive index: 1.559
- Liquid
Formula:
C₆H₅Br Boiling Pt: 156 °C Melting Pt: –31 °C Density: 1.491 g/mL Flash Pt: 51 °C (124 °F) |
MDL Number:
MFCD00000055 CAS Number: 108-86-1 EINECS: 203-623-8 UN: 2514 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...