Zirconium(IV) 2-ethylhexanoate ≥97%

Supplier: BeanTown Chemical

221275-100G 221275-25G
BT221275-100GEA 241 USD
BT221275-100G BT221275-25G
Zirconium(IV) 2-ethylhexanoate ≥97%
Zirconium(IV) 2-ethylhexanoate

CAS: 2233-42-3; EC No: 218-776-6; MDL No: MFCD00072684 Liquid; Molecular Formula: C32H60O8Zr ; MW: 664.04 Hygroscopic


  • Hygroscopic
  • Liquid

Formula: C₃₂H₆₀O₈Zr
MDL Number: MFCD00072684
CAS Number: 2233-42-3
EINECS: 218-776-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR