Tin ≥99.9999%, pieces

Supplier: BeanTown Chemical

221260-100G 221260-5G
BT221260-100GEA 698 USD
BT221260-100G BT221260-5G
Tin ≥99.9999%, pieces
Tin

CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862; RTECS: XP7320000 Solid; Molecular Formula: Sn; MW: 118.71 Melting Point: 231.9°; Boiling Point: 2270° Density (g/mL): 7.31


  • RTECS: XP7320000
  • Solid

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/mL
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

Specification Test Results

Assay 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR