Niobium diselenide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Niobium(IV) selenide

221220-10G 221220-50G
BT221220-10GEA 75 USD
BT221220-10G BT221220-50G
Niobium diselenide ≥99.5%
Niobium diselenide

CAS: 12034-77-4; EC No: 234-811-8; MDL No: MFCD00016228 UN No: UN3283; Haz Class: 6.1; Packing Group: III 5 Micron Powder; Molecular Formula: NbSe2; MW: 250.83 Melting Point: 1316° Density (g/mL): 6.3


  • 5 Micron Powder

Formula: NbSe₂
Melting Pt: >1316 °C
Density: 6.3 g/mL
MDL Number: MFCD00016228
CAS Number: 12034-77-4
EINECS: 234-811-8
UN: 3283
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR