D-Biotin-N-succinimidyl ester ≥98%

Supplier: BeanTown Chemical

Synonyms: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

221165-1G 221165-250MG
BT221165-1GEA 550.97 USD
BT221165-1G BT221165-250MG
D-Biotin-N-succinimidyl ester ≥98%
D-Biotin-N-succinimidyl ester

CAS: 35013-72-0; MDL No: MFCD00078531 Powder; Molecular Formula: C14H19N3O4S; MW: 341.38


  • Powder

Formula: C₁₄H₁₉N₃O₅S
MDL Number: MFCD00078531
CAS Number: 35013-72-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR