3-(3,4-Dihydroxyphenyl)-DL-alanine ≥96%
Supplier: BeanTown Chemical
Synonyms:
DL-β-3,4-Dihydroxyphenyl-Alanine, DL-DOPA, 3,4-Dihydroxy-DL-phenylalanine
221010-100G
221010-25G
221010-5G
BT221010-100GEA
556
USD
BT221010-100G
BT221010-25G
BT221010-5G
3-(3,4-Dihydroxyphenyl)-DL-alanine ≥96%
3-(3,4-Dihydroxyphenyl)-DL-alanine
CAS: 63-84-3; EC No: 200-566-0; MDL No: MFCD00063060; RTECS: AY5250000
Crystalline; Molecular Formula: C9H11NO4; MW: 197.19
Melting Point: 290° (decomposes)
- RTECS: AY5250000
- Crystalline
Formula:
C₉H₁₁NO₄ Melting Pt: 290 °C (dec.) |
MDL Number:
MFCD00063060 CAS Number: 63-84-3 EINECS: 200-566-0 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...