tert-Butyl isocyanide ≥98%
Supplier: BeanTown Chemical
221000-1G
221000-5G
BT221000-1GEA
51.68
USD
BT221000-1G
BT221000-5G
tert-Butyl isocyanide ≥98%
tert-Butyl isocyanide
CAS: 7188-38-7; EC No: 230-553-5; MDL No: MFCD00000002; RTECS: EQ7102500
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: (CH3)3CNC; MW: 83.13
Boiling Point: 90-92°; Flash point: -2°C (28°F)
Density (g/mL): 0.735; Refractive Index: 1.376
- RTECS: EQ7102500
- Refractive index: 1.376
- Liquid
Formula:
C₅H₉N Boiling Pt: 90…92 °C Density: 0.735 g/mL Flash Pt: –2 °C (28 °F) |
MDL Number:
MFCD00000002 CAS Number: 7188-38-7 EINECS: 230-553-5 UN: 1992 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...