(±)-3-Quinuclidinol ≥98%
Supplier: BeanTown Chemical
Synonyms:
(±)-Quinuclidin-3-ol, (±)-3-Hydroxyquinuclidine
220990-100G
220990-25G
220990-5G
BT220990-100GEA
370
USD
BT220990-100G
BT220990-25G
BT220990-5G
(±)-3-Quinuclidinol ≥98%
(±)-3-Quinuclidinol
CAS: 1619-34-7; EC No: 216-578-4; MDL No: MFCD00151326; RTECS: VD6191700
UN No: UN3263; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: C7H13NO; MW: 127.18
Melting Point: 220-223°
Air Sensitive
- RTECS: VD6191700
- Air Sensitive
- Powder
Formula:
C₇H₁₃NO Melting Pt: 220…223 °C |
MDL Number:
MFCD00151326 CAS Number: 1619-34-7 EINECS: 216-578-4 UN: 3263 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...