Diyttrium trioxide ≥99.9% (trace rare earth metals basis) aerosol refractory paint

Supplier: BeanTown Chemical

Synonyms: Yttrium oxide, Yttrium (III) oxide

220970-13OZ
BT220970-13OZEA 394 USD
BT220970-13OZ
Diyttrium trioxide ≥99.9% (trace rare earth metals basis) aerosol refractory paint
Diyttrium trioxide

CAS: 1314-36-9; MDL No: MFCD00011473 UN No: UN1950; Haz Class: 2.1 Aerosol Spray Can; Linear Formula: Y2O3; MW: 225.81 Air Sensitive


  • Air Sensitive
  • Aerosol Spray Can

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410
MDL Number: MFCD00011473
CAS Number: 1314-36-9
ADR: 2.1

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR