Molybdenum(V) isopropoxide 5% (w/v) in 2-propanol , 99.5% (trace metals basis)
Supplier: BeanTown Chemical
220885-100ML
220885-25ML
BT220885-100MLEA
529
USD
BT220885-100ML
BT220885-25ML
Molybdenum(V) isopropoxide 5% (w/v) in 2-propanol , 99.5% (trace metals basis)
Molybdenum(V) isopropoxide
CAS: 209733-38-0; MDL No: MFCD00210636
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C15H35MoO5 ; MW: 391.04
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₁₅H₃₅MoO₅ |
MDL Number:
MFCD00210636 CAS Number: 209733-38-0 ADR: 3,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...