2-Iodo-9,9-dimethylfluorene

Supplier: BeanTown Chemical

Synonyms: 2-Iodo-9,9-dimethyl-9H-fluorene

220875-25G 220875-5G
BT220875-25GEA 222 USD
BT220875-25G BT220875-5G
2-Iodo-9,9-dimethylfluorene
2-Iodo-9,9-dimethylfluorene

CAS: 144981-85-1 Solid; Molecular Formula: C15H13I; MW: 320.17 Melting Point: 63-65° Density (g/mL): 1.533


  • Solid

Formula: C₁₅H₁₃I
MW: 320.17 g/mol
Melting Pt: 63…65 °C
Density: 1.533 g/mL
MDL Number: MFCD12024272
CAS Number: 144981-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR