Calcium pyrophosphate ≥96%
Supplier: BeanTown Chemical
220855-1KG
220855-250G
BT220855-1KGEA
72
USD
BT220855-1KG
BT220855-250G
BT220855-5KG
Calcium pyrophosphate ≥96%
Calcium pyrophosphate
CAS: 7790-76-3; EC No: 232-221-5; MDL No: MFCD00015983
Powder; Linear Formula: Ca2P2O7; MW: 254.10
Melting Point: 1353°
Density (g/mL): 3.09
- Powder
Formula:
Ca₂O₇P₂ Melting Pt: 1353 °C Density: 3.09 g/mL |
MDL Number:
MFCD00015983 CAS Number: 7790-76-3 EINECS: 232-221-5 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...