1-Butanol ≥99.4% ACS

Supplier: BeanTown Chemical

Synonyms: Propylcarbinol, Methylolpropane, Butan-1-ol, Butyl alcohol, n-Butyl alcohol, Butyl hydroxide, 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxybutane, 1-Hydroxy butane, n-Butan-1-ol, Propyl carbinol, n-Butanol, n-Butyl Alcohol

220845-1L 220845-4L 220845-500ML
BT220845-1LEA 47.64 USD
BT220845-1L BT220845-4L BT220845-500ML
1-Butanol ≥99.4% ACS
1-Butanol

CAS: 71-36-3; EC No: 200-751-6; MDL No: MFCD00002964; RTECS: EO1400000 UN No: UN1120; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2)3OH; Molecular Formula: C4H10O; MW: 74.12 Melting Point: -90°; Boiling Point: 116-118°; Flash point: 35°C (95°F) Density (g/mL): 0.81; Refractive Index: 1.399 Hygroscopic


  • RTECS: EO1400000
  • Refractive index: 1.399
  • Hygroscopic
  • Liquid

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: -89
Density: 0.81 g/mL
Flash Pt: 35 °C (95 °F)
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.4% min.
Color (APHA) ≤10
Residue after evaporation ≤0.005%
Titrable acid ≤0.0008meq/g
Butyl ether ≤0.2%
Water ≤0.1%
Isobutanol ≤0.10%
Carbonyl compounds (as butyraldehyde) ≤0.01%
Specific gravity at 25°C 0.807-0.809


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR