Triethylammonium dihydrogentrifluoride

Supplier: BeanTown Chemical

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

220755-100G 220755-25G 220755-5G
BT220755-100GEA 219.3 USD
BT220755-100G BT220755-25G BT220755-5G
Triethylammonium dihydrogentrifluoride
Triethylammonium dihydrogentrifluoride

CAS: 73602-61-6; EC No: 277-550-5; MDL No: MFCD00043294 UN No: UN1790; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: (CH3CH2)3N·3HF; Molecular Formula: C6H15N·3HF; MW: 161.21 Melting Point: -29°; Boiling Point: 70°/15 mmHg; Flash point: 87°C (188°F) Density (g/mL): 0.989; Refractive Index: 1.392 Hygroscopic


  • Refractive index: 1.392
  • Hygroscopic
  • Liquid

Formula: C₆H₁₈F₃N
MW: 161.21
Boiling Pt: 70 °C/15 mmHg
Melting Pt: –29 °C
Density: 0.989 g/mL
Flash Pt: 87 °C (188 °F)
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 1790
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR