Barium aluminate, Technical Grade

Supplier: BeanTown Chemical

220740-100G 220740-500G
BT220740-100GEA 62 USD
BT220740-100G BT220740-500G
Barium aluminate, Technical Grade
Barium aluminate

CAS: 12004-04-5; EC No: 234-445-9; MDL No: MFCD00049716 UN No: UN1564; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: BaAl2O4 ; MW: 561.98


  • Powder

Formula: Al₂BaO₄
MW: 255.29 g/mol
MDL Number: MFCD00049716
CAS Number: 12004-04-5
EINECS: 234-445-9
UN: 1564
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR