Aluminium oxide, α-phase ≥99.95% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
ɑ-Tonerde, α-Alumina
220725-100G
220725-2KG
220725-500G
BT220725-100GEA
35
USD
BT220725-100G
BT220725-2KG
BT220725-500G
Aluminium oxide, α-phase ≥99.95% (trace metals basis)
Aluminium oxide, α-phase
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
0.35-0.49 Micron APS Powder, S.A. 7.0-9.0 m2/g ; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 0.35-0.49 Micron APS Powder, S.A. 7.0-9.0 m²/g
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...