Nitrilotris(methylenephosphonic acid) ≥97%
Supplier: BeanTown Chemical
Synonyms:
Tris(phosphonomethyl)amine, Nitrilotri(methylphosphonic acid)
220695-10G
220695-50G
BT220695-10GEA
93
USD
BT220695-10G
BT220695-50G
Nitrilotris(methylenephosphonic acid) ≥97%
Nitrilotris(methylenephosphonic acid)
CAS: 6419-19-8; EC No: 229-146-5; MDL No: MFCD00002138; RTECS: SZ9860000
Crystalline; Linear Formula: N[CH2PO(OH)2]3; Molecular Formula: C3H12NO9P3; MW: 299.05
Melting Point: 215° (decomposes)
Density (g/mL): 1.300
Hygroscopic
- RTECS: SZ9860000
- Hygroscopic
- Crystalline
Formula:
C₃H₁₂NO₉P₃ Melting Pt: 215 °C (dec.) Density: 1.300 g/mL |
MDL Number:
MFCD00002138 CAS Number: 6419-19-8 EINECS: 229-146-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...