1-Benzyl-4-hydroxypiperidine ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-Benzylpiperidin-4-ol
220650-100G
220650-25G
220650-5G
BT220650-100GEA
434
USD
BT220650-100G
BT220650-25G
BT220650-5G
1-Benzyl-4-hydroxypiperidine ≥97%
1-Benzyl-4-hydroxypiperidine
CAS: 4727-72-4; EC No: 225-226-9; MDL No: MFCD00006503
Solid; Molecular Formula: C12H18NO; MW: 191.27
Melting Point: 61-63°; Boiling Point: 127-128°/2 mmHg
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₁₂H₁₇NO Boiling Pt: 127…128 °C/2 mmHg Melting Pt: 61…63 °C |
MDL Number:
MFCD00006503 CAS Number: 4727-72-4 EINECS: 225-226-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...