Benzene-[D₆] ≥99.5% (Isotopic)
Supplier: BeanTown Chemical
CAS: 1076-43-3; EC No: 214-061-8; MDL No: MFCD00003010
UN No: UN1114; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6D6; MW: 84.15
Melting Point: 6.8°; Boiling Point: 79.1°; Flash point: -11°C (12°F)
Density (g/mL): 0.95; Refractive Index: 1.497
Moisture Sensitive
- Refractive index: 1.497
- Moisture Sensitive
- Liquid
Formula:
C₆D₆ Boiling Pt: 79.1 °C Melting Pt: 6.8 °C Density: 0.95 g/mL Flash Pt: –11 °C (12 °F) |
MDL Number:
MFCD00003010 CAS Number: 1076-43-3 EINECS: 214-061-8 UN: 1114 ADR: 3,II |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...