3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one ≥98%
Supplier: BeanTown Chemical
Synonyms:
Diethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphate, DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate
220565-25G
220565-5G
BT220565-25GEA
180
USD
BT220565-25G
BT220565-5G
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one ≥98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
CAS: 165534-43-0; MDL No: MFCD01236967
Solid; Molecular Formula: C11H14N3O5P ; MW: 299.22
Melting Point: 72-75°
- Solid
Formula:
C₁₁H₁₄N₃O₅P Melting Pt: 72…75 °C |
MDL Number:
MFCD01236967 CAS Number: 165534-43-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...