Lead(II) chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Lead dichloride

220540-2KG 220540-500G
BT220540-2KGEA 197 USD
BT220540-2KG BT220540-500G
Lead(II) chloride ≥99%
Lead (II) chloride

CAS: 7758-95-4; EC No: 231-845-5; MDL No: MFCD00011157 UN No: UN2291; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: PbCl2; MW: 278.10 Density (g/mL): 5.85


  • Crystalline/Powder

Formula: Cl₂Pb
Density: 5.85 g/mL
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR