Carbon nanotube multi-walled ≥95% <8 nm OD, 2-5 nm ID, Length 0.5-2 µm

Supplier: BeanTown Chemical

Synonyms: CNT

220510-1G 220510-25G 220510-5G
BT220510-1GEA 53 USD
BT220510-1G BT220510-25G BT220510-5G
Carbon nanotube multi-walled ≥95% <8 nm OD, 2-5 nm ID, Length 0.5-2 µm
Carbon nanotube

CAS: 308068-56-6; MDL No: MFCD06202029 UN No: UN1325; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: C; MW: 12.011


  • Powder

Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6
ADR: 4.1,III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR