Phosphoryl trichloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride
220470-1KG
220470-250G
BT220470-1KGEA
173.47
USD
BT220470-1KG
BT220470-250G
Phosphoryl trichloride ≥99%
Phosphoryl trichloride
CAS: 10025-87-3; EC No: 233-046-7; MDL No: MFCD00011443; RTECS: TH4897000
UN No: UN1810; Haz Class: 6.1 (8); Packing Group: I
Liquid; Linear Formula: POCl3; MW: 153.33
Melting Point: 1.25°; Boiling Point: 105.8°
Density (g/mL): 1.645; Refractive Index: 1.46
Moisture Sensitive
- RTECS: TH4897000
- Refractive index: 1.46
- Moisture Sensitive
- Liquid
Formula:
POCl₃ Boiling Pt: 105.8 °C Melting Pt: 1.25 °C Density: 1.645 g/mL |
MDL Number:
MFCD00011443 CAS Number: 10025-87-3 EINECS: 233-046-7 UN: 1810 ADR: 8,I |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...