2-Chlorothiazole ≥97%

Supplier: BeanTown Chemical

220460-1G 220460-5G
BT220460-1GEA 161 USD
BT220460-1G BT220460-5G
2-Chlorothiazole ≥97%
2-Chlorothiazole

CAS: 3034-52-4; EC No: 221-228-9; MDL No: MFCD00210701 Liquid; Molecular Formula: C3H2ClNS; MW: 119.57 Density (g/mL): 1.393; Refractive Index: 1.551


  • Refractive index: 1.551
  • Liquid

Formula: C₃H₂ClNS
Density: 1.393 g/mL
MDL Number: MFCD00210701
CAS Number: 3034-52-4
EINECS: 221-228-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR