4-Methylcyclohexanone ≥98%
Supplier: BeanTown Chemical
220435-100G
220435-25G
220435-500G
BT220435-100GEA
123.14
USD
BT220435-100G
BT220435-25G
BT220435-500G
4-Methylcyclohexanone ≥98%
4-Methylcyclohexanone
CAS: 589-92-4; EC No: 209-665-3; MDL No: MFCD00001643; RTECS: GW1750200
UN No: UN2297; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C7H12O; MW: 112.17
Boiling Point: 169-171°; Flash point: 48°C (118°F)
Density (g/mL): 0.914; Refractive Index: 1.444
- RTECS: GW1750200
- Refractive index: 1.444
- Liquid
Formula:
C₇H₁₂O Boiling Pt: 169…171 °C Density: 0.914 g/mL Flash Pt: 48 °C (118 °F) |
MDL Number:
MFCD00001643 CAS Number: 589-92-4 EINECS: 209-665-3 UN: 2297 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...