4-Methylcyclohexanone ≥98%

Supplier: BeanTown Chemical

220435-100G 220435-25G 220435-500G
BT220435-100GEA 123.14 USD
BT220435-100G BT220435-25G BT220435-500G
4-Methylcyclohexanone ≥98%
4-Methylcyclohexanone

CAS: 589-92-4; EC No: 209-665-3; MDL No: MFCD00001643; RTECS: GW1750200 UN No: UN2297; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C7H12O; MW: 112.17 Boiling Point: 169-171°; Flash point: 48°C (118°F) Density (g/mL): 0.914; Refractive Index: 1.444


  • RTECS: GW1750200
  • Refractive index: 1.444
  • Liquid

Formula: C₇H₁₂O
Boiling Pt: 169…171 °C
Density: 0.914 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: 3,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR