Polyethylene glycol 1,500

Supplier: BeanTown Chemical

Synonyms: PEG 1500, PEG 1,500

220390-1KG 220390-250G
BT220390-1KGEA 94.65 USD
BT220390-1KG BT220390-250G
Polyethylene glycol 1,500
Polyethylene glycol 1,500

CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD01779609; RTECS: TQ4030000 Powder; Linear Formula: H(OCH2CH2)nOH; MW ≈1,500 Melting Point: 44-48°; Flash point: 229°C (444°F) Density (g/mL): 1.210

Formula: H(OCH₂CH₂)nOH
MW: 1500 g/mol
Melting Pt: 44…48 °C
Density: 1.210 g/mL
Flash Pt: 229 °C (444 °F)
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR