Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
BOP-Cl
220370-1G
220370-25G
220370-5G
BT220370-1GEA
21
USD
BT220370-1G
BT220370-25G
BT220370-5G
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride ≥97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
CAS: 68641-49-6; MDL No: MFCD00010077; RTECS: SZ5871000
UN No: UN3261; Haz Class: 8; Packing Group: II
Crystalline; Molecular Formula: C6H8ClN2O5P; MW: 254.56
Melting Point: 191° (decomposes)
Moisture Sensitive
- RTECS: SZ5871000
- Moisture Sensitive
- Crystalline
Formula:
C₆H₈ClN₂O₅P Melting Pt: 191 °C (dec.) |
MDL Number:
MFCD00010077 CAS Number: 68641-49-6 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...