Polyvinylpyrrolidone, M.W. ∼ 8.000 (K16-18)
Supplier: BeanTown Chemical
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
220350-100G
220350-500G
BT220350-100GEA
86.21
USD
BT220350-100G
BT220350-500G
Polyvinylpyrrolidone, M.W. ∼ 8.000 (K16-18)
Polyvinylpyrrolidone
CAS: 9003-39-8; MDL No: MFCD00149016
Powder; Molecular Formula: (C6H9NO)n
Melting Point: 130°
- Powder
Formula:
(C₆H₉NO)n Melting Pt: 130 °C |
MDL Number:
MFCD01076626 CAS Number: 9003-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...