Aluminium oxide ≥99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Aluminium(III) oxide, Alumina C, Aluminum oxide
220265-2KG
220265-500G
BT220265-2KGEA
96
USD
BT220265-2KG
BT220265-500G
Aluminium oxide ≥99% (trace metals basis)
Aluminium oxide
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
Powder; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- Powder
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...