Zirconium(IV) oxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Zirconium dioxide, Zirconium Oxide
220080-100G
220080-500G
BT220080-100GEA
60
USD
BT220080-100G
BT220080-500G
Zirconium(IV) oxide ≥99.5%
Zirconium (IV) oxide
CAS: 1314-23-4; EC No: 215-227-2; MDL No: MFCD00011310; RTECS: ZH8800000
1 Micron APS Powder; Linear Formula: ZrO2; MW: 123.22
Melting Point: 2700°; Boiling Point: 5000°
Density (g/mL): 5.89
- RTECS: ZH8800000
- 1 Micron APS Powder
Formula:
ZrO₂ Boiling Pt: 5000 °C Melting Pt: 2700 °C Density: 5.89 g/mL |
MDL Number:
MFCD00011310 CAS Number: 1314-23-4 EINECS: 215-227-2 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...