Methotrexate
Supplier: BeanTown Chemical
Synonyms:
(S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
220060-100MG
220060-1G
BT220060-100MGEA
124
USD
BT220060-100MG
BT220060-1G
Methotrexate
Methotrexate
CAS: 59-05-2; EC No: 200-413-8; MDL No: MFCD00150847; RTECS: MA1225000
UN No: UN1544; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C20H22N8O5; MW: 454.44
Melting Point: 185-204°
- RTECS: MA1225000
- Powder
Formula:
C₂₀H₂₂N₈O₅ MW: 454.44 g/mol Melting Pt: 185…204 °C |
MDL Number:
MFCD00150847 CAS Number: 59-05-2 EINECS: 200-413-8 UN: 1544 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...