Disodium malonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium malonate dibasic, Malonic acid disodium salt

220050-100G 220050-25G
BT220050-100GEA 86.28 USD
BT220050-100G BT220050-25G
Disodium malonate ≥99%
Disodium malonate

CAS: 141-95-7; EC No: 205-514-0; MDL No: MFCD00002708; RTECS: OO1750000 Crystalline/Powder; Linear Formula: CH2(COONa)2; Molecular Formula: C3H2Na2O4; MW: 148.03


  • RTECS: OO1750000
  • Crystalline/Powder

Formula: C₃H₂Na₂O₄
MW: 148.03 g/mol
MDL Number: MFCD00002708
CAS Number: 141-95-7
EINECS: 205-514-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR