2,7-Dichlorofluorene ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,7-Dichloro-9H-fluorene

220045-1G 220045-5G
BT220045-1GEA 100 USD
BT220045-1G BT220045-5G
2,7-Dichlorofluorene ≥97%
2,7-Dichlorofluorene

CAS: 7012-16-0; MDL No: MFCD00032840 Powder; Molecular Formula: C13H8Cl2; MW: 235.11


  • Powder

Formula: C₁₃H₈Cl₂
MDL Number: MFCD00032840
CAS Number: 7012-16-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR