3-Bromoanisole ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-3-methoxybenzene

220025-100G 220025-25G 220025-500G
BT220025-100GEA 95 USD
BT220025-100G BT220025-25G BT220025-500G
3-Bromoanisole ≥98%
3-Bromoanisole

CAS: 2398-37-0; EC No: 219-264-5; MDL No: MFCD00000081 Liquid; Linear Formula: BrC6H4OCH3; Molecular Formula: C7H7BrO; MW: 187.03 Boiling Point: 210-211°; Flash point: 98°C (208°F) Density (g/mL): 1.477; Refractive Index: 1.563


  • Refractive index: 1.563
  • Liquid

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 211
Melting Pt: 2
Density: 1.477 g/mL
Flash Pt: 98 °C (208 °F)
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR