Polyethylene glycol 400
Supplier: BeanTown Chemical
Synonyms:
PEG 400
219970-1KG
219970-250G
BT219970-1KGEA
84.46
USD
BT219970-1KG
BT219970-250G
Polyethylene glycol 400
Polyethylene glycol 400
CAS: 25322-68-3; MDL No: MFCD01779601
Liquid; Linear Formula: H(OCH2CH2)nOH; MW ≈400
Melting Point: 4-8°; Flash point: 240°C (464°F)
Density (g/mL): 1.128; Refractive Index: 1.466
- Refractive index: 1.466
- Liquid
Formula:
H(OCH₂CH₂)nOH Melting Pt: 4…8 °C Density: 1.128 g/mL Flash Pt: 240 °C (464 °F) |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...