Cesium carbonate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
219905-25G
219905-5G
BT219905-25GEA
170.08
USD
BT219905-25G
BT219905-5G
Cesium carbonate ≥99.995% (trace metals basis)
Cesium carbonate
CAS: 534-17-8; EC No: 208-591-9; MDL No: MFCD00010957; RTECS: FK9400000
Powder; Linear Formula: Cs2CO3; MW: 325.82
Melting Point: 610° (decomposes)
Hygroscopic
- RTECS: FK9400000
- Hygroscopic
- Powder
Formula:
Cs₂CO₃ Melting Pt: 610 °C (dec.) |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...