2-Propanol ≥99.7% for spectrophotometry

Supplier: BeanTown Chemical

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

219850-1L 219850-4L 219850-4X1L
BT219850-1LEA 43.83 USD
BT219850-1L BT219850-4L BT219850-4X1L
2-Propanol ≥99.7% for spectrophotometry
2-Propanol

CAS: 67-63-0; EC No: 200-661-7; MDL No: MFCD00011674; RTECS: NT8050000 UN No: UN1219; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)2CHOH; Molecular Formula: C3H8O; MW: 60.10 Melting Point: -90°; Boiling Point: 82°; Flash point: 12°C (54°F) Density (g/mL): 0.785; Refractive Index: 1.377


  • RTECS: NT8050000
  • Refractive index: 1.377
  • Liquid

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.785 g/mL
Flash Pt: 12 °C (54 °F)
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR