2-Propanol ≥99.7% for spectrophotometry
Supplier: BeanTown Chemical
219850-1L
219850-4L
219850-4X1L
BT219850-1LEA
43.83
USD
BT219850-1L
BT219850-4L
BT219850-4X1L
2-Propanol ≥99.7% for spectrophotometry
2-Propanol
CAS: 67-63-0; EC No: 200-661-7; MDL No: MFCD00011674; RTECS: NT8050000
UN No: UN1219; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)2CHOH; Molecular Formula: C3H8O; MW: 60.10
Melting Point: -90°; Boiling Point: 82°; Flash point: 12°C (54°F)
Density (g/mL): 0.785; Refractive Index: 1.377
- RTECS: NT8050000
- Refractive index: 1.377
- Liquid
Formula:
(CH₃)₂CHOH Boiling Pt: 82 °C Melting Pt: –90 °C Density: 0.785 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II |
|
Specification Test Results
Assay | 99.7% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...