Bismuth triacetate ≥99%

Supplier: BeanTown Chemical

Synonyms: Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate

219805-250G 219805-50G
BT219805-250GEA 338 USD
BT219805-250G BT219805-50G
Bismuth triacetate ≥99%
Bismuth triacetate

CAS: 22306-37-2; EC No: 244-904-5; MDL No: MFCD00015629 Crystalline; Linear Formula: Bi(OOCCH3)3; Molecular Formula: C6H9BiO6; MW: 386.11 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: (H₃CCOO)₃Bi
MW: 386.11 g/mol
MDL Number: MFCD00015629
CAS Number: 22306-37-2
EINECS: 244-904-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR