Thallium(III) trifluoroacetate ≥95%

Supplier: BeanTown Chemical

Synonyms: Thallic trifluoroacetate

219730-1G 219730-5G
BT219730-1GEA 17 USD
BT219730-1G BT219730-5G
Thallium(III) trifluoroacetate ≥95%
Thallium (III) trifluoroacetate

CAS: 23586-53-0; EC No: 245-761-1; MDL No: MFCD00000414 UN No: UN1707; Haz Class: 6.1; Packing Group: II Solid; Linear Formula: Tl(OOCCF3)3; Molecular Formula: C6F9O6Tl; MW: 543.42 Melting Point: 213° (decomposes) Air Sensitive , Hygroscopic


  • Air Sensitive, Hygroscopic
  • Solid

Formula: C₆F₉O₆Tl
MW: 543.43 g/mol
Boiling Pt: 160
Melting Pt: 120.5 (dec.)
MDL Number: MFCD00000414
CAS Number: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1 II

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR