N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone
219650-10KG
219650-2.5KG
219650-500G
BT219650-10KGEA
301.83
USD
BT219650-10KG
BT219650-2.5KG
BT219650-500G
N-Methyl-2-pyrrolidone (NMP) ≥99%
N-Methyl-2-pyrrolidone (NMP)
CAS: 872-50-4; EC No: 212-828-1; MDL No: MFCD00003193; RTECS: UY5790000
Liquid; Molecular Formula: C5H9NO; MW: 99.13
Melting Point: -24°; Boiling Point: 202°; Flash point: 91°C (196°F)
Density (g/mL): 1.028; Refractive Index: 1.470
- RTECS: UY5790000
- Refractive index: 1.470
- Liquid
Formula:
C₅H₉NO MW: 99.13 g/mol Boiling Pt: 202 Melting Pt: -24 Density: 1.028 g/mL Flash Pt: 91 °C (196 °F) |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 13,06140 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...