4-Aminopyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Pyridin-4-amine, 4-Pyridylamine
219575-1KG
219575-250G
219575-50G
BT219575-1KGEA
790
USD
BT219575-1KG
BT219575-250G
BT219575-50G
4-Aminopyridine ≥98%
4-Aminopyridine
CAS: 504-24-5; EC No: 207-987-9; MDL No: MFCD00006439; RTECS: US1750000
UN No: UN2671; Haz Class: 6.1; Packing Group: II
Crystalline; Molecular Formula: C5H6N2; MW: 94.11
Melting Point: 155-158°; Boiling Point: 273°
- RTECS: US1750000
- Crystalline
Formula:
C₅H₆N₂ Boiling Pt: 273 °C Melting Pt: 155…158 °C |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...