1,3-Dichloropropane ≥95%

Supplier: BeanTown Chemical

Synonyms: Trimethylendichloride

219510-100G 219510-25G
BT219510-100GEA 262 USD
BT219510-100G BT219510-25G
1,3-Dichloropropane ≥95%
1,3-Dichloropropane

CAS: 142-28-9; EC No: 205-531-3; MDL No: MFCD00000999 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Linear Formula: Cl(CH2)3Cl; MW: 112.99 Melting Point: -99°; Boiling Point: 120-122°; Flash point: 36°C (97°F) Density (g/mL): 1.19; Refractive Index: 1.448


  • Refractive index: 1.448
  • Liquid

Formula: ClH₂CCH₂CH₂Cl
MW: 112.99
Boiling Pt: 120.4
Melting Pt: -99.5
Density: 1.19 g/mL
Flash Pt: 36 °C (97 °F)
MDL Number: MFCD00000999
CAS Number: 142-28-9
EINECS: 205-531-3
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR