D-Lactose monohydrate ACS
Supplier: BeanTown Chemical
CAS: 64044-51-5; EC No: 200-559-2; MDL No: MFCD00150747; RTECS: OD9625000
Crystalline; Molecular Formula: C12H22O11·H2O; MW: 360.32
Melting Point: 219° (decomposes)
Density (g/mL): 1.525; Optical Rotation: [α]22/D +52.4°, c = 4.5 in H2O
- RTECS: OD9625000
- Optical rotation: [α]22/D +52.4°, c = 4.5 in H2O
- Crystalline
Formula:
C₁₂H₂₂O₁₁·1H₂O Melting Pt: 219 °C (dec.) Density: 1.525 g/mL |
MDL Number:
MFCD00150747 CAS Number: 64044-51-5 EINECS: 200-559-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...