Ammonium tetrathiomolybdate ≥99.95%
Supplier: BeanTown Chemical
Synonyms:
Ammonium tetrathiomolybdate(VI)
219365-25G
219365-5G
BT219365-25GEA
419
USD
BT219365-25G
BT219365-5G
Ammonium tetrathiomolybdate ≥99.95%
Ammonium tetrathiomolybdate
CAS: 15060-55-6; MDL No: MFCD00136013; RTECS: QA4668250
Crystalline; Linear Formula: (NH4)2MoS4; MW: 260.28
Melting Point:
- RTECS: QA4668250
- Hygroscopic
- Crystalline
Formula:
H₁₀MoN₂S₄ Melting Pt: >300 °C |
MDL Number:
MFCD00136013 CAS Number: 15060-55-6 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...