6-Chlorooxindole ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Chloro-2-oxindole
219275-100G
219275-25G
219275-5G
BT219275-100GEA
548
USD
BT219275-100G
BT219275-25G
BT219275-5G
6-Chlorooxindole ≥98%
6-Chlorooxindole
CAS: 56341-37-8; MDL No: MFCD00209962
Crystalline/Powder; Molecular Formula: C8H6ClNO ; MW: 167.59
Melting Point: 195-199°
- Crystalline/Powder
Formula:
C₈H₆ClNO Melting Pt: 195…199 °C |
MDL Number:
MFCD00209962 CAS Number: 56341-37-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...