2-Bromothiophene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Thienyl bromide

219195-1KG 219195-250G 219195-50G
BT219195-1KGEA 455 USD
BT219195-1KG BT219195-250G BT219195-50G
2-Bromothiophene ≥98%
2-Bromothiophene

CAS: 1003-09-4; EC No: 213-699-4; MDL No: MFCD00005417 UN No: UN2929; Haz Class: 6.1 (3); Packing Group: II Liquid; Molecular Formula: C4H3BrS ; MW: 163.04 Boiling Point: 149-151°; Flash point: 52°C (126°F) Density (g/mL): 1.684; Refractive Index: 1.586 Light Sensitive


  • Refractive index: 1.586
  • Light Sensitive
  • Liquid

Formula: C₄H₃BrS
Boiling Pt: 149…151 °C
Density: 1.684 g/mL
Flash Pt: 52 °C (126 °F)
MDL Number: MFCD00005417
CAS Number: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR