Potassium methanesulfonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Potassiummethanesulfonate, Potassiummethanesulphonate, Methanesulphonic acid potassium salt, Methanesulfonic acid potassium salt

219185-100G 219185-25G 219185-500G
BT219185-100GEA 130 USD
BT219185-100G BT219185-25G BT219185-500G
Potassium methanesulfonate ≥99%
Potassium methanesulphonate

CAS: 2386-56-3; EC No: 219-202-7; MDL No: MFCD00070544; RTECS: PB2675000 Powder/Lump; Linear Formula: CH3SO3K; MW: 134.19 Hygroscopic


  • RTECS: PB2675000
  • Hygroscopic
  • Powder/Lump

Formula: CH₃KO₃S
MW: 134.2 g/mol
MDL Number: MFCD00070544
CAS Number: 2386-56-3
EINECS: 219-202-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR