Cobalt ≥99.5% (trace metals basis), powder -325 mesh
Supplier: BeanTown Chemical
219080-100G
219080-500G
BT219080-100GEA
126.63
USD
BT219080-100G
BT219080-500G
Cobalt ≥99.5% (trace metals basis), powder -325 mesh
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- RTECS: GF8750000
- Powder
Formula:
Co Boiling Pt: 2900 °C Melting Pt: 1495 °C Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...